4-Phenylazophenol 97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-(Phenyldiazenyl)phenol
417360250
417361000
ACRO417360250EA
40.4
GBP
ACRO417360250
ACRO417361000
4-Phenylazophenol 97%
4-Phenylazophenol
Formula:
C₁₂H₁₀N₂O MW: 198.23 g/mol Melting Pt: 150…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002330 CAS Number: 1689-82-3 EINECS: 216-880-6 |
Specification Test Results
Appearance | Brown Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...