5-Iodo-1,2,3-trimethoxybenzene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

711535-1G 711535-5G
SIAL711535-1GEA 128 GBP
SIAL711535-1G SIAL711535-5G
5-Iodo-1,2,3-trimethoxybenzene, Sigma-Aldrich®
5-Iodo-1,2,3-trimethoxybenzene
Formula: C₉H₁₁IO₃
MW: 294.09 g/mol
MDL Number: MFCD01318153
CAS Number: 25245-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR