3,5-Diiodo-2-methoxyphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-8041-1G
BB-8041-250M
COBBBB-8041-1GEA
0
GBP
COBBBB-8041-1G
COBBBB-8041-250M
3,5-Diiodo-2-methoxyphenylboronic acid
3,5-Diiodo-2-methoxyphenylboronic acid
Formula:
C₇H₇BI₂O₃ |
MDL Number:
MFCD08457647 CAS Number: 1072951-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...