3-(Trimethoxysilyl)propyl methacrylate
Supplier: abcr
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
γ-methacryloxypropyl-trimethoxysilane, 3-(Methacryloyloxy)propyltrimethoxysilane
SIM6487.4.0100
AB109006.0100
AB109006-25KG
ABCRSIM6487.4.0100EA
0
GBP
ABCRSIM6487.4.0100
ABCRAB109006.0100
ABCRAB109006-25KG
3-(Trimethoxysilyl)propyl methacrylate
3-(Trimethoxysilyl)propyl methacrylate
Formula:
C₁₀H₂₀O₅Si |
MDL Number:
MFCD00008593 CAS Number: 2530-85-0 EINECS: 219-785-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...