(Methylamino)acetaldehyde dimethylacetal, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,2-Dimethoxy-N-methylethanamine

M-Clarity™ quality level = MQ100

M28006-25G
SIALM28006-25GEA 65.3 GBP
SIALM28006-25G
(Methylamino)acetaldehyde dimethylacetal, Sigma-Aldrich®
(Methylamino)acetaldehyde dimethylacetal
Formula: C₅H₁₃NO₂
MW: 119.16 g/mol
MDL Number: MFCD00008485
CAS Number: 122-07-6
EINECS: 204-520-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR