(±)-2-Pyrrolidinone-5-carboxylic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Oxopyrrolidine-2-carboxylic acid, DL-2-Pyrrolidone-5-carboxylic acid, DL-Pyroglutamic acid

M-Clarity™ quality level = MQ200

292915-100G 292915-5G
SIAL292915-100GEA 94.3 GBP
SIAL292915-100G SIAL292915-5G
(±)-2-Pyrrolidinone-5-carboxylic acid, Sigma-Aldrich®
(±)-2-Pyrrolidinone-5-carboxylic acid
Formula: C₅H₇NO₃
MW: 129.12 g/mol
MDL Number: MFCD00064322
CAS Number: 149-87-1
EINECS: 205-748-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR