1-(4-Fluoro-2-nitrophenyl)pyrrolidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF1141.1 017161-1G
CHMPCFF1141.1EA 0 GBP
CHMPCFF1141.1 FLUO017161-1G
1-(4-Fluoro-2-nitrophenyl)pyrrolidine
1-(4-Fluoro-2-nitrophenyl)pyrrolidine
Formula: C₁₀H₁₁ClN₂O₂
MW: 210.20 g/mol
MDL Number: MFCD00798372
CAS Number: 778-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR