1-(3-Chloro-4-fluorophenyl)pyrrolidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6445-5G
HC-6445-1G
COBBHC-6445-5GEA
0
GBP
COBBHC-6445-5G
COBBHC-6445-1G
1-(3-Chloro-4-fluorophenyl)pyrrolidine
1-(3-Chloro-4-fluorophenyl)pyrrolidine
Formula:
C₁₀H₁₁ClFN |
MDL Number:
MFCD09878399 CAS Number: 1000339-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...