1-(3-Chloro-2-methylphenyl)pyrrolidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6490-5G
HC-6490-1G
COBBHC-6490-5GEA
0
GBP
COBBHC-6490-5G
COBBHC-6490-1G
1-(3-Chloro-2-methylphenyl)pyrrolidine
1-(3-Chloro-2-methylphenyl)pyrrolidine
Formula:
C₁₁H₁₄ClN |
MDL Number:
MFCD09972165 CAS Number: 1020253-09-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...