1-(5-Bromo-2-nitrophenyl)pyrrolidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1282-5G AN-1282-25G AN-1282-1G
COBBAN-1282-5GEA 0 GBP
COBBAN-1282-5G COBBAN-1282-25G COBBAN-1282-1G
1-(5-Bromo-2-nitrophenyl)pyrrolidine
1-(5-Bromo-2-nitrophenyl)pyrrolidine
Formula: C₁₀H₁₁BrN₂O₂
MDL Number: MFCD10699604
CAS Number: 1033201-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR