1-(3-Bromo-5-methylphenyl)pyrrolidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-1004-5G OT-1004-1G
COBBOT-1004-5GEA 0 GBP
COBBOT-1004-5G COBBOT-1004-1G
1-(3-Bromo-5-methylphenyl)pyrrolidine
1-(3-Bromo-5-methylphenyl)pyrrolidine
Formula: C₁₁H₁₄BrN
MDL Number: MFCD13195737
CAS Number: 1199773-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR