1,3-Dichloro-6-methylisoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2545-5G HC-2545-1G
COBBHC-2545-5GEA 0 GBP
COBBHC-2545-5G COBBHC-2545-1G
1,3-Dichloro-6-methylisoquinoline
1,3-Dichloro-6-methylisoquinoline
Formula: C₁₀H₇Cl₂N
MW: 212.0800 g/mol
MDL Number: MFCD11112095
CAS Number: 21902-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR