Triethyl phosphonoacetate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001326-100G 001326-25G
FLUO001326-100GEA 0 GBP
FLUO001326-100G FLUO001326-25G
Triethyl phosphonoacetate
Triethyl phosphonoacetate
Formula: C₈H₁₇O₅P
MDL Number: MFCD00009177
CAS Number: 867-13-0
EINECS: 212-757-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR