8-Bromoisoquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6020-5G
HC-6020-1G
COBBHC-6020-5GEA
0
GBP
COBBHC-6020-5G
COBBHC-6020-1G
8-Bromoisoquinoline
8-Bromoisoquinoline
Formula:
C₉H₆BrN MW: 208.0600 g/mol |
MDL Number:
MFCD04973298 CAS Number: 63927-22-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...