5,8-Dibromoisoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6032-1G HC-6032-5G
COBBHC-6032-1GEA 0 GBP
COBBHC-6032-1G COBBHC-6032-5G
5,8-Dibromoisoquinoline
5,8-Dibromoisoquinoline
Formula: C₉H₅Br₂N
MW: 286.95 g/mol
MDL Number: MFCD03109888
CAS Number: 81045-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR