3-Acetamido-5-bromobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4115-5G CA-4115-1G
COBBCA-4115-5GEA 0 GBP
COBBCA-4115-5G COBBCA-4115-1G
3-Acetamido-5-bromobenzoic acid
3-Acetamido-5-bromobenzoic acid
Formula: C₉H₈BrNO₃
MDL Number: MFCD01098204
CAS Number: 78238-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR