3-Chloro-2-methoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Chloro-2-methoxyphenyl)boronic acid

BB-5212-5G BB-5212-1G BB-5212-250M
COBBBB-5212-5GEA 0 GBP
COBBBB-5212-5G COBBBB-5212-1G COBBBB-5212-250M
3-Chloro-2-methoxyphenylboronic acid
3-Chloro-2-methoxyphenylboronic acid
Formula: C₇H₈BClO₃
MW: 186.4006 g/mol
MDL Number: MFCD08277278
CAS Number: 179898-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR