Glycylglycine (Gly-Gly)

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

51460.02 51460.03
SERA51460.02EA 0 GBP
SERA51460.02 SERA51460.03
Glycylglycine (Gly-Gly)
Proteins and Peptides

  • EINECS: 209-127-8
  • GHS symbol and signal word: GHS07 and warning

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR