Glycylglycine (Gly-Gly)
Supplier: BACHEM BIOCHEMICA
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4030664.0025
4030664.0100
BACE4030664.0025EA
0
GBP
BACE4030664.0025
BACE4030664.0100
Glycylglycine (Gly-Gly)
Proteins and Peptides
- EINECS: 209-127-8
- GHS symbol and signal word: GHS07 and warning
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...