Benzyl carbamate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-1205-100G
OT-1205-25G
COBBOT-1205-100GEA
0
GBP
COBBOT-1205-100G
COBBOT-1205-25G
Benzyl carbamate
Benzyl carbamate
Formula:
C₈H₉NO₂ |
MDL Number:
MFCD00007965 CAS Number: 621-84-1 EINECS: 210-710-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...