3-Chloro-4-fluorophenylacetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005670-1G 005670-5G
FLUO005670-1GEA 0 GBP
FLUO005670-1G FLUO005670-5G
3-Chloro-4-fluorophenylacetonitrile
3-Chloro-4-fluorophenylacetonitrile
Formula: C₈H₅ClFN
MW: 169.5834 g/mol
MDL Number: MFCD01631553
CAS Number: 658-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR