3,5-Dibromo-2-methylthiophene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1697-1G
HI-1697-5G
COBBHI-1697-1GEA
0
GBP
COBBHI-1697-1G
COBBHI-1697-5G
3,5-Dibromo-2-methylthiophene
3,5-Dibromo-2-methylthiophene
Formula:
C₅H₄Br₂S MW: 255.9583 g/mol |
MDL Number:
MFCD07781197 CAS Number: 29421-73-6 EINECS: 249-617-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...