3,3'-Iminobis(propylamine), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,3'-Iminobis(propylamine), 3,3'-Iminodipropylamine, 3,3'-Diaminodipropylamine, Dipropylenetriamine

M-Clarity™ quality level = MQ200

I1006-100G-A I1006-500G-A
SIALI1006-100G-AEA 35.9 GBP
SIALI1006-100G-A SIALI1006-500G-A
3,3'-Iminobis(propylamine), Sigma-Aldrich®
3,3'-Iminobis(propylamine)
Formula: C₆H₁₇N₃
MDL Number: MFCD00008214
CAS Number: 56-18-8
EINECS: 200-261-2
UN: 2269
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR