1-(2,3-Dimethylphenyl)pyrrolidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6390-5G HC-6390-1G
COBBHC-6390-5GEA 0 GBP
COBBHC-6390-5G COBBHC-6390-1G
1-(2,3-Dimethylphenyl)pyrrolidine
1-(2,3-Dimethylphenyl)pyrrolidine
Formula: C₁₂H₁₇N
MDL Number: MFCD09878389
CAS Number: 957065-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR