3-Fluoro-5-methoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Fluoro-5-methoxybenzeneboronic acid

BB-2775-5G BB-2775-25G BB-2775-1G
COBBBB-2775-5GEA 0 GBP
COBBBB-2775-5G COBBBB-2775-25G COBBBB-2775-1G
3-Fluoro-5-methoxyphenylboronic acid
3-Fluoro-5-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD07363780
CAS Number: 609807-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR