3-Fluoro-2-methoxyphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-5110-5G
BB-5110-1G
COBBBB-5110-5GEA
0
GBP
COBBBB-5110-5G
COBBBB-5110-1G
3-Fluoro-2-methoxyphenylboronic acid
3-Fluoro-2-methoxyphenylboronic acid
Formula:
C₇H₈BFO₃ MW: 169.9460 g/mol |
MDL Number:
MFCD03095355 CAS Number: 762287-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...