3,4-Difluoro-5-methoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3,4-Difluoro-5-methoxyphenyl)boronic acid

BB-9054-5G BB-9054-1G
COBBBB-9054-5GEA 0 GBP
COBBBB-9054-5G COBBBB-9054-1G
3,4-Difluoro-5-methoxyphenylboronic acid
3,4-Difluoro-5-methoxyphenylboronic acid
Formula: C₇H₇BF₂O₃
MW: 187.940 g/mol
MDL Number: MFCD09907960
CAS Number: 925910-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR