2-Fluoro-6-methoxyphenylboronic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-6-methoxybenzeneboronic acid

M-Clarity™ quality level = MQ100

512761-1G 512761-5G
SIAL512761-1GEA 44.6 GBP
SIAL512761-1G SIAL512761-5G
2-Fluoro-6-methoxyphenylboronic acid, Sigma-Aldrich®
2-Fluoro-6-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169.95 g/mol
MDL Number: MFCD02179483
CAS Number: 78495-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR