5-Isoquinolinol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Hydroxyisoquinoline

M-Clarity™ quality level = MQ100

H33207-25G H33207-5G
SIALH33207-25GEA 693 GBP
SIALH33207-25G SIALH33207-5G
5-Isoquinolinol, Sigma-Aldrich®
5-Isoquinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD00006906
CAS Number: 2439-04-5
EINECS: 219-456-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR