3-Chloro-2-hydrazinopyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7086-1G
PY-7086-5G
COBBPY-7086-1GEA
0
GBP
COBBPY-7086-1G
COBBPY-7086-5G
3-Chloro-2-hydrazinopyridine
3-Chloro-2-hydrazinopyridine
Formula:
C₅H₆ClN₃ |
MDL Number:
MFCD01936479 CAS Number: 22841-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...