2-Butoxy-5-methylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8024-1G BB-8024-250M
COBBBB-8024-1GEA 0 GBP
COBBBB-8024-1G COBBBB-8024-250M
2-Butoxy-5-methylphenylboronic acid
2-Butoxy-5-methylphenylboronic acid
Formula: C₁₁H₁₇BO₃
MW: 208.06 g/mol
MDL Number: MFCD04974068
CAS Number: 480438-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR