cis-Aconitic acid
Supplier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(Z)-Prop-1-ene-1,2,3-tricarboxylic acid, (1Z)-Prop-1-ene-1,2,3-tricarboxylic acid, (Z)-Aconitic acid
10640.02
10640.03
SERA10640.02EA
0
GBP
SERA10640.02
SERA10640.03
cis-Aconitic acid
cis-Aconitic acid
Formula:
C₆H₆O₆ MW: 174.11 g/mol |
MDL Number:
MFCD00063184 CAS Number: 585-84-2 EINECS: 209-564-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...