N-(2-Pyridyl)thiourea
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2-Pyridyl)thiourea, 1-(2-Pyridyl)-2-thiourea, 1-(Pyridin-2-yl)thiourea
018986-10G
018986-1G
018986-5G
FLUO018986-10GEA
0
GBP
FLUO018986-10G
FLUO018986-1G
FLUO018986-5G
N-(2-Pyridyl)thiourea
N-(2-Pyridyl)thiourea
Formula:
C₆H₇N₃S MW: 153.21 g/mol |
MDL Number:
MFCD00041227 CAS Number: 14294-11-2 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...