Nonadecanoic acid methyl ester, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Nonadecanoic acid methyll ester, Methyl Nonadecanoate

M-Clarity™ quality level = MQ200

N5377-1G N5377-10G N5377-5G
SIALN5377-1GEA 170 GBP
SIALN5377-1G SIALN5377-10G SIALN5377-5G
Nonadecanoic acid methyl ester, Sigma-Aldrich®
Nonadecanoic acid methyl ester
Formula: C₂₀H₄₀O₂
MDL Number: MFCD00009011
CAS Number: 1731-94-8
EINECS: 217-056-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR