2-Pyrimidinecarbonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Cyanopyrimidine
019246-10G
019246-50G
FLUO019246-10GEA
0
GBP
FLUO019246-10G
FLUO019246-50G
2-Pyrimidinecarbonitrile
2-Pyrimidinecarbonitrile
Formula:
C₅H₃N₃ |
MDL Number:
MFCD00160513 CAS Number: 14080-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...