3-Chloro-6-hydrazinopyridazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Chloro-6-hydrazinylpyridazine

HC-6415-5G HC-6415-25G HC-6415-1G
COBBHC-6415-5GEA 0 GBP
COBBHC-6415-5G COBBHC-6415-25G COBBHC-6415-1G
3-Chloro-6-hydrazinopyridazine
3-Chloro-6-hydrazinopyridazine
Formula: C₄H₅ClN₄
MW: 144.56 g/mol
MDL Number: MFCD00051740
CAS Number: 17284-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR