N-(Benzyloxycarbonyloxy)succinimide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: CBZ-OSu (Z-OSu)

011620-1KG 011620-25G
FLUO011620-1KGEA 0 GBP
FLUO011620-1KG FLUO011620-25G
N-(Benzyloxycarbonyloxy)succinimide
N-(Benzyloxycarbonyloxy)succinimide
Formula: C₁₂H₁₁NO₅
MDL Number: MFCD00005513
CAS Number: 13139-17-8
EINECS: 236-075-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR