5,6,7,8-Tetrahydro-1-naphthol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7720-5G OS-7720-25G
COBBOS-7720-5GEA 0 GBP
COBBOS-7720-5G COBBOS-7720-25G
5,6,7,8-Tetrahydro-1-naphthol
5,6,7,8-Tetrahydro-1-naphthol
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00001734
CAS Number: 529-35-1
EINECS: 208-461-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR