Potassium dicyanoaurate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

379867-1G 379867-250MG
SIAL379867-1GEA 513 GBP
SIAL379867-1G SIAL379867-250MG
Potassium dicyanoaurate, Sigma-Aldrich®
Potassium dicyanoaurate
Formula: C₂AuKN₂
MDL Number: MFCD00011414
CAS Number: 13967-50-5
EINECS: 237-748-4
UN: 1588
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR