2-Butoxy-5-chlorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Butoxy-5-chlorophenyl)boronic acid

BB-8122-1G BB-8122-250M
COBBBB-8122-1GEA 0 GBP
COBBBB-8122-1G COBBBB-8122-250M
2-Butoxy-5-chlorophenylboronic acid
2-Butoxy-5-chlorophenylboronic acid
Formula: C₁₀H₁₄BClO₃
MW: 228.48 g/mol
MDL Number: MFCD08276746
CAS Number: 352534-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR