5-Chloro-2-fluorobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013958-1G 013958-5G
FLUO013958-1GEA 0 GBP
FLUO013958-1G FLUO013958-5G
5-Chloro-2-fluorobenzaldehyde
5-Chloro-2-fluorobenzaldehyde
Formula: C₇H₄ClFO
MDL Number: MFCD03094518
CAS Number: 96515-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR