3-Butoxy-2,4,6-trifluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-8095-1G BB-8095-250M
COBBBB-8095-1GEA 0 GBP
COBBBB-8095-1G COBBBB-8095-250M
3-Butoxy-2,4,6-trifluorophenylboronic acid
3-Butoxy-2,4,6-trifluorophenylboronic acid
Formula: C₁₀H₁₂BF₃O₃
MW: 248.01 g/mol
MDL Number: MFCD07784387
CAS Number: 871126-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR