Malonyl dichloride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
M1601-100G
M1601-25G
M1601-5G
SIALM1601-100GEA
373
GBP
SIALM1601-100G
SIALM1601-25G
SIALM1601-5G
Malonyl dichloride, Sigma-Aldrich®
Malonyl dichloride
Formula:
C₃H₂Cl₂O₂ MW: 140.95 g/mol |
MDL Number:
MFCD00000735 CAS Number: 1663-67-8 EINECS: 216-772-9 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...