2-Chloro-4-propylpyrimidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6167-1G HC-6167-5G
COBBHC-6167-1GEA 0 GBP
COBBHC-6167-1G COBBHC-6167-5G
2-Chloro-4-propylpyrimidine
2-Chloro-4-propylpyrimidine
Formula: C₇H₉ClN₂
MDL Number: MFCD09800976
CAS Number: 111196-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR