3-Butoxy-4-methoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5969-5G BB-5969-1G
COBBBB-5969-5GEA 189.93 GBP
COBBBB-5969-5G COBBBB-5969-1G
3-Butoxy-4-methoxyphenylboronic acid
3-Butoxy-4-methoxyphenylboronic acid
Formula: C₁₁H₁₇BO₄
MDL Number: MFCD13195649
CAS Number: 1217501-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR