Methyl 4-aminonicotinate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013940-1G 013940-5G 013940-25G
FLUO013940-1GEA 0 GBP
FLUO013940-1G FLUO013940-5G FLUO013940-25G
Methyl 4-aminonicotinate
Methyl 4-aminonicotinate
Formula: C₇H₈N₂O₂
MDL Number: MFCD00955996
CAS Number: 16135-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR