4-Chloro-2-(trifluoromethyl)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Chloro-2-(trifluoromethyl)benzeneboronic acid, (4-Chloro-2-(trifluoromethyl)phenyl)metaboric acid, Dihydroxy(4-chloro-2-(trifluoromethyl)phenyl)borane
BB-2265-5G
BB-2265-25G
BB-2265-1G
COBBBB-2265-5GEA
0
GBP
COBBBB-2265-5G
COBBBB-2265-25G
COBBBB-2265-1G
4-Chloro-2-(trifluoromethyl)phenylboronic acid
4-Chloro-2-(trifluoromethyl)phenylboronic acid
Formula:
C₇H₅BClF₃O₂ |
MDL Number:
MFCD04973086 CAS Number: 313545-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...