3-(Bromomethyl)phenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Bromomethylphenylboronic acid, 3-(Bromomethyl)benzeneboronic acid, Dihydroxy(3-(bromomethyl)phenyl)borane, (3-(Bromomethyl)phenyl)metaboric acid
BB-2468-5G
BB-2468-25G
BB-2468-1G
COBBBB-2468-5GEA
0
GBP
COBBBB-2468-5G
COBBBB-2468-25G
COBBBB-2468-1G
3-(Bromomethyl)phenylboronic acid
3-(Bromomethyl)phenylboronic acid
Formula:
C₇H₈BBrO₂ MW: 214.85 g/mol |
MDL Number:
MFCD01632207 CAS Number: 51323-43-4 UN: 3261 ADR: 8,III |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...