2-(Trifluoromethoxy)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Trifluoromethoxyphenylboronic acid, 2-(Trifluoromethoxy)benzeneboronic acid, (2-(Trifluoromethoxy)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethoxy)phenyl)borane

BB-2650-1G BB-2650-5G BB-2650-25G BB-2650-100G
COBBBB-2650-1GEA 0 GBP
COBBBB-2650-1G COBBBB-2650-5G COBBBB-2650-25G COBBBB-2650-100G
2-(Trifluoromethoxy)phenylboronic acid
2-(Trifluoromethoxy)phenylboronic acid
Formula: C₇H₆BF₃O₃
MW: 205.93 g/mol
MDL Number: MFCD01320763
CAS Number: 175676-65-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR