2,4,5-Trichloropyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20412G100 AS20412G25
ACTIAS20412G100EA 0 GBP
ACTIAS20412G100 ACTIAS20412G25
2,4,5-Trichloropyrimidine
2,4,5-Trichloropyrimidine
Formula: C₄HCl₃N₂
Boiling Pt: 94 °C (12 mmHg)
MDL Number: MFCD03788200
CAS Number: 5750-76-5
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR